Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338016.1 | NP_001326496.1 | 13 - 14 | 4505 | 81 | |
NM_112135.4 | NP_187904.1 | 13 - 14 | 4523 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |