Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112163.6 | NP_187929.4 | 5 - 6 | 745 | 157 | |
GenBank mRNA | AY063883.1 | - | 5 - 6 | 704 | 157 | |
AY142588.1 | - | 5 - 6 | 673 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |