Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112210.3 | NP_187973.1 | 4 - 5 | 492 | 76 | |
GenBank mRNA | BT003947.1 | - | 4 - 5 | 376 | 76 | |
BT005042.1 | - | 3 - 4 | 286 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |