Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112210.3 | NP_187973.1 | 8 - 9 | 1202 | 89 | |
GenBank mRNA | BT003947.1 | - | 8 - 9 | 1086 | 89 | |
BT005042.1 | - | 7 - 8 | 996 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |