Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP90D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338068.1 | NP_001327080.1 | 1 - 2 | 334 | 83 | |
NM_112223.3 | NP_566462.1 | 1 - 2 | 334 | 83 | ||
GenBank mRNA | AB066286.1 | - | 1 - 2 | 250 | 83 | |
BT004084.1 | - | 1 - 2 | 334 | 83 | ||
BT005093.1 | - | 1 - 2 | 250 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |