Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338075.1 | NP_001319544.1 | 5 - 6 | 520 | 84 | |
NM_112230.3 | NP_187993.2 | 4 - 5 | 401 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |