Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATBFRUCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202952.1 | NP_001189881.1 | 3 - 4 | 1269 | 77 | |
NM_001338079.1 | NP_001327313.1 | 2 - 3 | 1716 | 77 | ||
NM_001338080.1 | NP_001327314.1 | 3 - 4 | 1590 | 77 | ||
NM_001338081.1 | NP_001327315.1 | 3 - 4 | 1116 | 77 | ||
NM_112232.4 | NP_566464.1 | 4 - 5 | 1280 | 77 | ||
GenBank mRNA | AY045776.1 | - | 4 - 5 | 1269 | 77 | |
AY079422.1 | - | 4 - 5 | 1242 | 77 | ||
X74514.1 | - | 1261 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |