Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATBFRUCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202952.1 | NP_001189881.1 | 5 - 6 | 1600 | 98 | |
NM_001338079.1 | NP_001327313.1 | 4 - 5 | 2047 | 98 | ||
NM_001338080.1 | NP_001327314.1 | 5 - 6 | 1921 | 98 | ||
NM_001338081.1 | NP_001327315.1 | 5 - 6 | 1447 | 98 | ||
NM_112232.4 | NP_566464.1 | 6 - 7 | 1611 | 98 | ||
GenBank mRNA | AY045776.1 | - | 6 - 7 | 1600 | 98 | |
AY079422.1 | - | 6 - 7 | 1573 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |