Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112241.4 | NP_188003.1 | 20 - 21 | 2249 | 90 | |
NM_202579.1 | NP_974308.1 | 20 - 21 | 2158 | 90 | ||
GenBank mRNA | AY092988.1 | - | 20 - 21 | 2093 | 90 | |
AY128801.1 | - | 19 - 20 | 2003 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |