Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112241.4 | NP_188003.1 | 5 - 6 | 632 | 67 | |
NM_202579.1 | NP_974308.1 | 5 - 6 | 541 | 67 | ||
GenBank mRNA | AY092988.1 | - | 5 - 6 | 476 | 67 | |
AY128801.1 | - | 4 - 5 | 386 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |