Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112299.4 | NP_566486.1 | 7 - 8 | 2180 | 95 | |
GenBank mRNA | AF302673.1 | - | 7 - 8 | 1875 | 95 | |
BT021128.1 | - | 7 - 8 | 1896 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |