Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202964.2 | NP_001189893.1 | 16 - 17 | 2183 | 69 | |
NM_112317.3 | NP_188075.2 | 16 - 17 | 2342 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |