Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202964.2 | NP_001189893.1 | 29 - 30 | 3412 | 99 | |
NM_001338124.1 | NP_001326811.1 | 28 - 29 | 3562 | 99 | ||
NM_112317.3 | NP_188075.2 | 28 - 29 | 3649 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |