Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP72A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112322.3 | NP_188079.1 | 4 - 5 | 1356 | 111 | |
GenBank mRNA | AY072303.1 | - | 4 - 5 | 1214 | 111 | |
BT003388.1 | - | 4 - 5 | 1115 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |