Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP72A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112329.4 | NP_188086.1 | 3 - 4 | 889 | 89 | |
GenBank mRNA | AF367347.1 | - | 3 - 4 | 798 | 89 | |
BT001008.1 | - | 3 - 4 | 741 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |