Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112333.3 | NP_188090.2 | 8 - 9 | 1689 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |