Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119316.5 | NP_194895.1 | 8 - 9 | 1252 | 129 | |
GenBank mRNA | AY142532.1 | - | 8 - 9 | 1145 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |