Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342110.1 | NP_001328539.1 | 3 - 4 | 646 | 122 | |
NM_001342111.1 | NP_001328538.1 | 3 - 4 | 690 | 122 | ||
NM_119324.4 | NP_194903.2 | 3 - 4 | 515 | 122 | ||
GenBank mRNA | AY080658.1 | - | 3 - 4 | 468 | 122 | |
AY133761.1 | - | 2 - 3 | 207 | 122 | ||
EU214909.1 | - | 2 - 3 | 207 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |