Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342110.1 | NP_001328539.1 | 6 - 7 | 990 | 67 | |
NM_001342111.1 | NP_001328538.1 | 6 - 7 | 1034 | 67 | ||
NM_119324.4 | NP_194903.2 | 6 - 7 | 859 | 67 | ||
GenBank mRNA | AY080658.1 | - | 6 - 7 | 812 | 67 | |
AY133761.1 | - | 5 - 6 | 551 | 67 | ||
EU214909.1 | - | 5 - 6 | 551 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |