Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342115.1 | NP_001320110.1 | 2 - 3 | 690 | 71 | |
NM_119329.4 | NP_567882.1 | 4 - 5 | 1158 | 71 | ||
GenBank mRNA | AF428421.1 | - | 4 - 5 | 732 | 71 | |
AK227401.1 | - | 4 - 5 | 732 | 71 | ||
AY125557.1 | - | 4 - 5 | 639 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |