Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203955.1 | NP_001190884.1 | 19 - 20 | 2077 | 87 | |
NM_001342121.1 | NP_001328314.1 | 17 - 18 | 2151 | 87 | ||
NM_119341.2 | NP_194918.2 | 17 - 18 | 2151 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |