Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342125.1 | NP_001328629.1 | 1 - 2 | 671 | 97 | |
NM_119356.4 | NP_194932.1 | 1 - 2 | 676 | 97 | ||
GenBank mRNA | AF306661.1 | - | 1 - 2 | 556 | 97 | |
BT026033.1 | - | 1 - 2 | 538 | 97 | ||
D43962.1 | - | 1 - 2 | 582 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |