Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036691.1 | NP_001031768.1 | 7 - 8 | 1137 | 93 | |
NM_001125620.1 | NP_001119092.1 | 7 - 8 | 1137 | 93 | ||
NM_119370.5 | NP_194945.3 | 7 - 8 | 1187 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |