Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125626.1 | NP_001119098.1 | 5 - 6 | 780 | 100 | |
NM_119404.3 | NP_567895.1 | 5 - 6 | 993 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |