Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036696.1 | NP_001031773.1 | 9 - 10 | 1617 | 117 | |
NM_119417.4 | NP_194991.1 | 9 - 10 | 1819 | 117 | ||
NM_202936.1 | NP_974665.1 | 9 - 10 | 1626 | 117 | ||
GenBank mRNA | AY052233.1 | - | 9 - 10 | 1617 | 117 | |
BT003093.1 | - | 9 - 10 | 1569 | 117 | ||
U81239.1 | - | 9 - 10 | 1574 | 117 | ||
Z83202.1 | - | 1615 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |