Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036697.1 | NP_001031774.1 | 3 - 4 | 346 | 102 | |
NM_119418.3 | NP_194992.1 | 2 - 3 | 582 | 102 | ||
NM_202937.1 | NP_974666.1 | 2 - 3 | 472 | 102 | ||
GenBank mRNA | BT029776.1 | - | 2 - 3 | 204 | 102 | |
D45355.1 | - | 2 - 3 | 263 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |