Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119430.5 | NP_567906.1 | 1 - 2 | 436 | 65 | |
GenBank mRNA | AF302188.1 | - | 1 - 2 | 239 | 65 | |
AK226552.1 | - | 1 - 2 | 306 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |