Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMMA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119448.4 | NP_195020.1 | 3 - 4 | 916 | 136 | |
GenBank mRNA | AF370160.1 | - | 3 - 4 | 628 | 136 | |
AF424619.1 | - | 3 - 4 | 626 | 136 | ||
AY059104.1 | - | 3 - 4 | 554 | 136 | ||
AY133531.1 | - | 3 - 4 | 554 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |