Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XI-I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203968.1 | NP_001190897.1 | 9 - 10 | 1396 | 77 | |
NM_001203969.1 | NP_001190898.1 | 9 - 10 | 1396 | 77 | ||
NM_001342203.1 | NP_001328260.1 | 9 - 10 | 1418 | 77 | ||
NM_119474.5 | NP_195046.3 | 9 - 10 | 1430 | 77 | ||
GenBank mRNA | AK228969.1 | - | 9 - 10 | 1417 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |