Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119499.4 | NP_195071.1 | 1 - 2 | 331 | 111 | |
GenBank mRNA | AY142517.1 | - | 1 - 2 | 160 | 111 | |
AY519611.1 | - | 1 - 2 | 160 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |