Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342219.1 | NP_001329920.1 | 1 - 2 | 523 | 85 | |
NM_119508.4 | NP_195079.2 | 1 - 2 | 547 | 85 | ||
GenBank mRNA | AY099556.1 | - | 1 - 2 | 377 | 85 | |
BT010357.1 | - | 1 - 2 | 292 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |