Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342233.1 | NP_001329220.1 | 6 - 7 | 1095 | 81 | |
NM_119537.7 | NP_567936.5 | 6 - 7 | 915 | 81 | ||
GenBank mRNA | AY057657.1 | - | 6 - 7 | 901 | 81 | |
AY070065.1 | - | 6 - 7 | 903 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |