Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342244.1 | NP_001320129.1 | 2 - 3 | 686 | 73 | |
NM_119556.3 | NP_567945.1 | 2 - 3 | 858 | 73 | ||
GenBank mRNA | AY054624.1 | - | 2 - 3 | 284 | 73 | |
AY081538.1 | - | 2 - 3 | 239 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |