Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119564.5 | NP_567950.1 | 3 - 4 | 1064 | 109 | |
GenBank mRNA | AF059510.1 | - | 3 - 4 | 871 | 109 | |
BT000384.1 | - | 3 - 4 | 793 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |