Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125640.1 | NP_001119112.1 | 15 - 16 | 2600 | 143 | |
NM_001342250.1 | NP_001329149.1 | 16 - 17 | 2827 | 143 | ||
NM_001342251.1 | NP_001329150.1 | 13 - 14 | 2852 | 143 | ||
NM_001342252.1 | NP_001329148.1 | 14 - 15 | 2700 | 143 | ||
NM_119567.4 | NP_195132.3 | 15 - 16 | 2754 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |