Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125640.1 | NP_001119112.1 | 19 - 20 | 2964 | 118 | |
NM_001342250.1 | NP_001329149.1 | 20 - 21 | 3238 | 118 | ||
NM_001342251.1 | NP_001329150.1 | 17 - 18 | 3263 | 118 | ||
NM_001342252.1 | NP_001329148.1 | 18 - 19 | 3111 | 118 | ||
NM_119567.4 | NP_195132.3 | 19 - 20 | 3165 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |