Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119573.5 | NP_567952.1 | 6 - 7 | 874 | 75 | |
GenBank mRNA | AF360331.1 | - | 6 - 7 | 642 | 75 | |
AY056339.1 | - | 6 - 7 | 587 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |