Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342263.1 | NP_001329319.1 | 2 - 3 | 393 | 71 | |
NM_001342264.1 | NP_001329320.1 | 1 - 2 | 336 | 71 | ||
NM_119588.6 | NP_567962.1 | 2 - 3 | 532 | 71 | ||
NM_202950.3 | NP_974679.1 | 2 - 3 | 340 | 71 | ||
GenBank mRNA | AY054633.1 | - | 2 - 3 | 183 | 71 | |
AY081532.1 | - | 2 - 3 | 159 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |