Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342263.1 | NP_001329319.1 | 8 - 9 | 1489 | 72 | |
NM_001342264.1 | NP_001329320.1 | 8 - 9 | 1344 | 72 | ||
NM_119588.6 | NP_567962.1 | 9 - 10 | 1540 | 72 | ||
GenBank mRNA | AY054633.1 | - | 9 - 10 | 1191 | 72 | |
AY081532.1 | - | 9 - 10 | 1167 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |