Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119600.4 | NP_567965.1 | 5 - 6 | 966 | 90 | |
GenBank mRNA | AY042877.1 | - | 5 - 6 | 936 | 90 | |
AY081454.1 | - | 5 - 6 | 860 | 90 | ||
AY168881.1 | - | 5 - 6 | 884 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |