Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119600.4 | NP_567965.1 | 8 - 9 | 1348 | 73 | |
GenBank mRNA | AY042877.1 | - | 8 - 9 | 1318 | 73 | |
AY081454.1 | - | 8 - 9 | 1242 | 73 | ||
AY168881.1 | - | 8 - 9 | 1266 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |