Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342287.1 | NP_001329504.1 | 6 - 7 | 1312 | 104 | |
NM_119614.6 | NP_195175.1 | 6 - 7 | 1271 | 104 | ||
GenBank mRNA | AB014759.1 | - | 6 - 7 | 1148 | 104 | |
BT029161.1 | - | 6 - 7 | 1093 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |