Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125644.1 | NP_001119116.1 | 4 - 5 | 1344 | 107 | |
NM_001342295.1 | NP_001328814.1 | 3 - 4 | 1941 | 107 | ||
NM_001342296.1 | NP_001328815.1 | 4 - 5 | 1416 | 107 | ||
NM_119627.4 | NP_195187.1 | 4 - 5 | 1729 | 107 | ||
GenBank mRNA | BT012291.1 | - | 3 - 4 | 1077 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |