Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT4G34840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342303.1 | NP_001329682.1 | 7 - 8 | 613 | 127 | |
NM_001342305.1 | NP_001329685.1 | 6 - 7 | 713 | 127 | ||
NM_119650.4 | NP_195210.2 | 7 - 8 | 646 | 127 | ||
GenBank mRNA | AK229730.1 | - | 7 - 8 | 619 | 127 | |
BT010195.1 | - | 7 - 8 | 569 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |