Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119666.4 | NP_195226.1 | 2 - 3 | 417 | 75 | |
GenBank mRNA | AY065143.1 | - | 2 - 3 | 325 | 75 | |
AY081646.1 | - | 2 - 3 | 237 | 75 | ||
AY086162.1 | - | 2 - 3 | 351 | 75 | ||
U69138.1 | - | 2 - 3 | 363 | 75 | ||
X98003.1 | - | 307 | 75 | |||
X98276.1 | - | 243 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |