Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR1;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334809.1 | NP_001321366.1 | 7 - 8 | 1812 | 70 | |
NM_001334810.1 | NP_001321365.1 | 7 - 8 | 1224 | 70 | ||
NM_001334811.1 | NP_001321364.1 | 7 - 8 | 1398 | 70 | ||
NM_001334812.1 | NP_001321363.1 | 6 - 7 | 1204 | 70 | ||
NM_001334813.1 | NP_001321362.1 | 5 - 6 | 678 | 70 | ||
NM_106449.3 | NP_565166.1 | 7 - 8 | 1398 | 70 | ||
NM_179568.2 | NP_849899.1 | 6 - 7 | 1230 | 70 | ||
GenBank mRNA | AB042322.2 | - | 7 - 8 | 992 | 70 | |
AK227518.1 | - | 6 - 7 | 977 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |