Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR1;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334809.1 | NP_001321366.1 | 8 - 9 | 1930 | 73 | |
NM_001334810.1 | NP_001321365.1 | 8 - 9 | 1342 | 73 | ||
NM_001334811.1 | NP_001321364.1 | 8 - 9 | 1516 | 73 | ||
NM_001334812.1 | NP_001321363.1 | 7 - 8 | 1322 | 73 | ||
NM_001334813.1 | NP_001321362.1 | 6 - 7 | 796 | 73 | ||
NM_106449.3 | NP_565166.1 | 8 - 9 | 1516 | 73 | ||
NM_179568.2 | NP_849899.1 | 7 - 8 | 1348 | 73 | ||
GenBank mRNA | AB042322.2 | - | 8 - 9 | 1110 | 73 | |
AK227518.1 | - | 7 - 8 | 1095 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |