Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106603.3 | NP_178073.1 | 5 - 6 | 1198 | 82 | |
NM_179576.3 | NP_849907.1 | 6 - 7 | 1215 | 82 | ||
GenBank mRNA | AF247560.1 | - | 5 - 6 | 1043 | 82 | |
AF348582.1 | - | 5 - 6 | 1009 | 82 | ||
AY062863.1 | - | 6 - 7 | 1116 | 82 | ||
BT008486.1 | - | 5 - 6 | 1009 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |