Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106616.5 | NP_178085.1 | 1 - 2 | 937 | 122 | |
NM_179577.1 | NP_849908.1 | 2 - 3 | 807 | 122 | ||
GenBank mRNA | AY078961.1 | - | 1 - 2 | 933 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |