Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179622.4 | NP_849953.2 | 5 - 6 | 582 | 92 | |
GenBank mRNA | AY337762.1 | - | 5 - 6 | 565 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |